accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 594-56-9

IUPAC Name: 2,3,3-trimethylbut-1-ene
Synonyms: 2,3,3-trimethyl-1-butene
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: AUYRUAVCWOAHQN-UHFFFAOYSA-N
SMILES: CC(=C)C(C)(C)C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)