accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 594-89-8

IUPAC Name: 1,1,1,2,2,3,3-heptachloropropane
Synonyms: 1,1,1,2,2,3,3-heptachloropropane
Molecular Weight (g/mol): 285.19
Molecular Formula: C3HCl7
InChi Key: YFIIENAGGCUHIQ-UHFFFAOYSA-N
SMILES: ClC(Cl)C(Cl)(Cl)C(Cl)(Cl)Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results