accessibility menu, dialog, popup

UserName

Viewing profile as user:
IOI

CAS RN 5943-11-3

IUPAC Name: 4,12-diiodo-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Synonyms: 4,12-diiodo-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular Weight (g/mol): 419.99
Molecular Formula: C12H6I2O
InChi Key: VKTLGHDNPWVFSY-UHFFFAOYSA-N
SMILES: IC1=CC2=C(OC3=C2C=C(I)C=C3)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results