accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNH

CAS RN 5946-39-4

IUPAC Name: 2,3,4,5-tetrahydro-1H-1,4-benzodiazepine
Synonyms: 2,3,4,5-tetrahydro-1H-1,4-benzodiazepine
Molecular Weight (g/mol): 148.21
Molecular Formula: C9H12N2
InChi Key: MLXBHOCKBUILHN-UHFFFAOYSA-N
SMILES: C1CNC2=CC=CC=C2CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results