accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)(S)H3C(R)HOOHO

CAS RN 5947-49-9

IUPAC Name: (1S,4aS,10aR)-6-hydroxy-1,4a-dimethyl-1,2,3,4,4a,9,10,10a-octahydrophenanthrene-1-carboxylic acid
Synonyms: podocarpic acid
Molecular Weight (g/mol): 274.36
Molecular Formula: C17H22O3
InChi Key: VJILEYKNALCDDV-OIISXLGYSA-N
SMILES: C[C@@]1(CCC[C@@]2(C)[C@H]1CCC1=CC=C(O)C=C21)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results