CAS RN 59480-92-1
IUPAC Name:
(2R,5S)-2,5-dimethyl-2,5-dihydro-1H-pyrrol-1-ium
Synonyms:
(2R,5S)-2,5-dimethyl-2,5-dihydro-1H-pyrrol-1-ium
Molecular Weight (g/mol):
98.17
Molecular Formula:
C6H12N
InChi Key:
TXQDHQBSNAJSHQ-OLQVQODUSA-O
SMILES:
C[C@H]1[NH2+][C@@H](C)C=C1
1
–
1
of
1
results
1
2,5-Dimethyl-3-pyrroline, 75%, mixture of cis and trans, Thermo Scientific™
CAS: 59480-92-1 Molecular Formula: C6H12N Molecular Weight (g/mol): 98.17 MDL Number: MFCD00005215 InChI Key: TXQDHQBSNAJSHQ-OLQVQODUSA-O PubChem CID: 101066 IUPAC Name: 2,5-dimethyl-2,5-dihydro-1H-pyrrole SMILES: C[C@H]1[NH2+][C@@H](C)C=C1