accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CNH2OHO

CAS RN 595-40-4

IUPAC Name: 2-amino-2-methylbutanoic acid
Synonyms: isovaline
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: GCHPUFAZSONQIV-UHFFFAOYNA-N
SMILES: CCC(C)(N)C(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)