accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 595-91-5

IUPAC Name: triphenylacetic acid
Synonyms: triphenylacetic acid
Molecular Weight (g/mol): 288.35
Molecular Formula: C20H16O2
InChi Key: DCYGAPKNVCQNOE-UHFFFAOYSA-N
SMILES: OC(=O)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)