accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3H3C(Z)

CAS RN 5953-76-4

IUPAC Name: methyl (2Z)-2-methylbut-2-enoate
Synonyms: methyl tiglate
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: YYJWBYNQJLBIGS-PLNGDYQASA-N
SMILES: COC(=O)C(\C)=C/C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results