accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59554-39-1

IUPAC Name: 2-methoxy-4-methylbenzene-1-sulfonamide
Synonyms: 2-methoxy-4-methylbenzenesulfonamide
Molecular Weight (g/mol): 201.24
Molecular Formula: C8H11NO3S
InChi Key: GQUFSEOISDLWMN-UHFFFAOYSA-N
SMILES: COC1=C(C=CC(C)=C1)S(N)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results