accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5957-90-4

IUPAC Name: 4-chloro-N-{2-[4-(4-chlorophenyl)-4H,5H,6H,7H-thieno[3,2-c]pyridin-5-yl]-2-oxoethyl}-N-(prop-2-en-1-yl)benzene-1-sulfonamide
Synonyms: 4-chloro-N-{2-[4-(4-chlorophenyl)-4H,6H,7H-thieno[3,2-c]pyridin-5-yl]-2-oxoethyl}-N-(prop-2-en-1-yl)benzenesulfonamide
Molecular Weight (g/mol): 521.47
Molecular Formula: C24H22Cl2N2O3S2
InChi Key: JDROFLIARIUQMO-UHFFFAOYNA-N
SMILES: ClC1=CC=C(C=C1)C1N(CCC2=C1C=CS2)C(=O)CN(CC=C)S(=O)(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results