accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5959-95-5

IUPAC Name: (2R)-2-amino-4-carbamoylbutanoic acid
Synonyms: D-glutamine D-glutaminsaeure-5-amid D-glutamin
Molecular Weight (g/mol): 146.15
Molecular Formula: C5H10N2O3
InChi Key: ZDXPYRJPNDTMRX-GSVOUGTGSA-N
SMILES: N[C@H](CCC(N)=O)C(O)=O
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (4)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)