accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 596-43-0

IUPAC Name: (bromodiphenylmethyl)benzene
Synonyms: (bromodiphenylmethyl)benzene
Molecular Weight (g/mol): 323.23
Molecular Formula: C19H15Br
InChi Key: NZHXEWZGTQSYJM-UHFFFAOYSA-N
SMILES: BrC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)