accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHH2NH2N

CAS RN 59649-56-8

IUPAC Name: 2,3-diaminophenol
Synonyms: 2,3-diaminophenol
Molecular Weight (g/mol): 124.14
Molecular Formula: C6H8N2O
InChi Key: PCAXITAPTVOLGL-UHFFFAOYSA-N
SMILES: NC1=C(N)C(O)=CC=C1
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)