accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOSCH3

CAS RN 59662-65-6

IUPAC Name: 1-methyl-4-(methylsulfanyl)methanesulfonylbenzene
Synonyms: 1-methyl-4-(methylsulfanyl)methanesulfonylbenzene
Molecular Weight (g/mol): 216.31
Molecular Formula: C9H12O2S2
InChi Key: XAARLLNXZJTFPQ-UHFFFAOYSA-N
SMILES: CSCS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results