accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SCH3NOONCH3SNCH3OONCH3SCH3(E)(E)

CAS RN 59669-26-0

IUPAC Name: (E)-[1-(methylsulfanyl)ethylidene]amino N-methyl-N-{[(3E)-3-methyl-6-oxo-5-oxa-2-thia-4,7-diazaoct-3-en-7-yl]sulfanyl}carbamate
Synonyms: thiodicarb
Molecular Weight (g/mol): 354.46
Molecular Formula: C10H18N4O4S3
InChi Key: XDOTVMNBCQVZKG-MKICQXMISA-N
SMILES: CS\C(C)=N\OC(=O)N(C)SN(C)C(=O)O\N=C(/C)SC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results