accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHH3CH3C

CAS RN 597-49-9

IUPAC Name: 3-ethylpentan-3-ol
Synonyms: 3-ethyl-3-pentanol
Molecular Weight (g/mol): 116.2
Molecular Formula: C7H16O
InChi Key: XKIRHOWVQWCYBT-UHFFFAOYSA-N
SMILES: CCC(O)(CC)CC
Boiling Point 
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)