accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 597-50-2

IUPAC Name: (diethylphosphoryl)ethane
Synonyms: (diethylphosphoryl)ethane
Molecular Weight (g/mol): 134.16
Molecular Formula: C6H15OP
InChi Key: ZSSWXNPRLJLCDU-UHFFFAOYSA-N
SMILES: CCP(=O)(CC)CC
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results