CAS RN 597-76-2
IUPAC Name:
3-ethylhexan-3-ol
Synonyms:
3-ethyl-3-hexanol
Molecular Weight (g/mol):
130.23
Molecular Formula:
C8H18O
InChi Key:
WNDLTOTUHMHNOC-UHFFFAOYSA-N
SMILES:
CCCC(O)(CC)CC
1
–
1
of
1
results
1
Promotions Available
3-Ethyl-3-hexanol, 98%, Thermo Scientific Chemicals
3-Ethyl-3-hexanol, 98%, Thermo Scientific Chemicals
CAS: 597-76-2 Molecular Formula: C8H18O Molecular Weight (g/mol): 130.23 MDL Number: MFCD00039616 InChI Key: WNDLTOTUHMHNOC-UHFFFAOYSA-N Synonym: 3-ethyl-3-hexanol,3-hexanol, 3-ethyl,1,1-diethyl-1-butanol,diethyl n-propyl carbinol,acmc-1ak6h PubChem CID: 69008 IUPAC Name: 3-ethylhexan-3-ol SMILES: CCCC(O)(CC)CC