accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 597-96-6

IUPAC Name: 3-methylhexan-3-ol
Synonyms: 3-methyl-3-hexanol
Molecular Weight (g/mol): 116.2
Molecular Formula: C7H16O
InChi Key: KYWJZCSJMOILIZ-UHFFFAOYNA-N
SMILES: CCCC(C)(O)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results