accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59734-92-8

IUPAC Name: 1-bromo-2-cyclohexylbenzene
Synonyms: 1-bromo-2-cyclohexylbenzene
Molecular Weight (g/mol): 239.16
Molecular Formula: C12H15Br
InChi Key: BXMDAGGQRIDSPA-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1C1CCCCC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results