accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 597554-76-2

IUPAC Name: 9,9,18,18,27,27-hexamethylheptacyclo[18.7.0.0²,¹⁰.0³,⁸.0¹¹,¹⁹.0¹²,¹⁷.0²¹,²⁶]heptacosa-1(20),2(10),3,5,7,11(19),12,14,16,21,23,25-dodecaene
Synonyms: 9,9,18,18,27,27-hexamethylheptacyclo[18.7.0.0²,¹⁰.0³,⁸.0¹¹,¹⁹.0¹²,¹⁷.0²¹,²⁶]heptacosa-1(20),2(10),3,5,7,11(19),12,14,16,21,23,25-dodecaene
Molecular Weight (g/mol): 426.6
Molecular Formula: C33H30
InChi Key: IZSZGRWXUIXAQK-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC=CC=C2C2=C1C1=C(C3=C2C(C)(C)C2=CC=CC=C32)C(C)(C)C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results