accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59777-95-6

IUPAC Name: 2-(pyridin-3-yl)-1,3-thiazolidine-4-carboxylic acid
Synonyms: 2-(pyridin-3-yl)-1,3-thiazolidine-4-carboxylic acid
Molecular Weight (g/mol): 210.25
Molecular Formula: C9H10N2O2S
InChi Key: FSNGLHIMQHWTNF-UHFFFAOYNA-N
SMILES: OC(=O)C1CSC(N1)C1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results