accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5978-08-5

IUPAC Name: 2-(3-chloropropyl)-2-methyl-1,3-dioxolane
Synonyms: 2-(3-chloropropyl)-2-methyl-1,3-dioxolane
Molecular Weight (g/mol): 164.63
Molecular Formula: C7H13ClO2
InChi Key: OFERIRWCHSOJJT-UHFFFAOYSA-N
SMILES: CC1(CCCCl)OCCO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results