CAS RN 5979-01-1
CAS RN 5979-01-1
IUPAC Name:
2-amino-1,4,5,6-tetrahydropteridine-4,6-dione hydrate
Synonyms:
xanthopterin hydrate
Molecular Weight (g/mol):
197.15
Molecular Formula:
C6H7N5O3
InChi Key:
GXYCFNCAIXIUMR-UHFFFAOYSA-N
SMILES:
O.NC1=NC(=O)C2=C(N1)N=CC(=O)N2
1
–
1
of
1
results
1
Xanthopterin Hydrate 98.0+%, TCI America™

CAS: 5979-01-1 Molecular Formula: C6H7N5O3 Molecular Weight (g/mol): 197.15 MDL Number: MFCD00151241 InChI Key: GXYCFNCAIXIUMR-UHFFFAOYSA-N Synonym: 2-Amino-4,6-pteridinediol PubChem CID: 2723796 IUPAC Name: 2-amino-1,5-dihydropteridine-4,6-dione;hydrate SMILES: C1=NC2=C(C(=O)N=C(N2)N)NC1=O.O