accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59798-30-0

IUPAC Name: sodium 6-{2-[(3-methanesulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetamido}-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Synonyms: sodium 6-[2-(3-methanesulfonyl-2-oxoimidazolidine-1-carbonylamino)-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol): 561.56
Molecular Formula: C21H24N5NaO8S2
InChi Key: GTGQRSIMEUWHPA-UHFFFAOYNA-M
SMILES: [Na+].CC1(C)SC2C(NC(=O)C(NC(=O)N3CCN(C3=O)S(C)(=O)=O)C3=CC=CC=C3)C(=O)N2C1C([O-])=O
Color 
  • (2)
Quantity 
  • (1)
  • (1)
Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results