CAS RN 598-01-6
IUPAC Name:
4-methylheptan-4-ol
Synonyms:
4-heptanol, 4-methyl-
Molecular Weight (g/mol):
130.23
Molecular Formula:
C8H18O
InChi Key:
IQXKGRKRIRMQCQ-UHFFFAOYSA-N
SMILES:
CCCC(C)(O)CCC
1
–
1
of
1
results
1
4-Methyl-4-heptanol 98.0+%, TCI America™
CAS: 598-01-6 Molecular Formula: C8H18O Molecular Weight (g/mol): 130.231 MDL Number: MFCD00021840 InChI Key: IQXKGRKRIRMQCQ-UHFFFAOYSA-N PubChem CID: 136394 IUPAC Name: 4-methylheptan-4-ol SMILES: CCCC(C)(CCC)O