accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2NH2NCH3NH2NH2NOHSOHOO

CAS RN 598-12-9

IUPAC Name: bis(N''-methylguanidine); sulfuric acid
Synonyms: bis(methyl-guanidine); sulfuric acid
Molecular Weight (g/mol): 244.27
Molecular Formula: C4H16N6O4S
InChi Key: WTYJBGQERCCGAD-UHFFFAOYSA-N
SMILES: CN=C(N)N.CN=C(N)N.OS(O)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results