accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBrO

CAS RN 598-22-1

IUPAC Name: propanoyl bromide
Synonyms: propionyl bromide
Molecular Weight (g/mol): 136.98
Molecular Formula: C3H5BrO
InChi Key: RIBFXMJCUYXJDZ-UHFFFAOYSA-N
SMILES: CCC(Br)=O
Boiling Point 
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (2)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (4)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)