accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH2C

CAS RN 598-25-4

IUPAC Name: 3-methylbuta-1,2-diene
Synonyms: 3-methyl-1,2-butadiene
Molecular Weight (g/mol): 68.12
Molecular Formula: C5H8
InChi Key: PAKGDPSCXSUALC-UHFFFAOYSA-N
SMILES: CC(C)=C=C
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)