accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 598-32-3

IUPAC Name: but-3-en-2-ol
Synonyms: 3-buten-2-ol
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: MKUWVMRNQOOSAT-UHFFFAOYNA-N
SMILES: CC(O)C=C
Boiling Point 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (7)
Quantity 
  • (2)
  • (2)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (7)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)