accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 598-55-0

IUPAC Name: methyl carbamate
Synonyms: methyl carbamate
Molecular Weight (g/mol): 75.07
Molecular Formula: C2H5NO2
InChi Key: GTCAXTIRRLKXRU-UHFFFAOYSA-N
SMILES: COC(N)=O
Percent Purity 
  • (2)
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (2)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)