accessibility menu, dialog, popup

UserName

Viewing profile as user:
MnOOO

CAS RN 598-62-9

IUPAC Name: manganese(2+) carbonate
Synonyms: manganese carbonate manganese(II) carbonate
Molecular Weight (g/mol): 114.95
Molecular Formula: CMnO3
InChi Key: XMWCXZJXESXBBY-UHFFFAOYSA-L
SMILES: [Mn++].[O-]C([O-])=O
Molecular Weight (g/mol) 
  • (7)
  • (8)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (13)
Percent Purity 
  • (2)
  • (6)
Grade 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (6)
  • (2)
  • (6)
  • (7)
  • (7)
  • (8)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (13)
  • (2)
  • (6)
  • (6)