accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 598-77-6

IUPAC Name: 1,1,2-trichloropropane
Synonyms: 1,1,2-trichloropropane
Molecular Weight (g/mol): 147.42
Molecular Formula: C3H5Cl3
InChi Key: GRSQYISVQKPZCW-UHFFFAOYNA-N
SMILES: CC(Cl)C(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results