accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 598-99-2

IUPAC Name: methyl 2,2,2-trichloroacetate
Synonyms: methyl trichloroacetate
Molecular Weight (g/mol): 177.41
Molecular Formula: C3H3Cl3O2
InChi Key: VHFUHRXYRYWELT-UHFFFAOYSA-N
SMILES: COC(=O)C(Cl)(Cl)Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results