CAS RN 59813-05-7
IUPAC Name:
dihydrogen piperazine dibromide
Synonyms:
dihydrogen piperazine dibromide
Molecular Weight (g/mol):
247.96
Molecular Formula:
C4H12Br2N2
InChi Key:
UXWKNNJFYZFNDI-UHFFFAOYSA-N
SMILES:
[H+].[H+].[Br-].[Br-].C1CNCCN1
1
–
1
of
1
results
1
Piperazine Dihydrobromide 98.0+%, TCI America™
CAS: 59813-05-7 Molecular Formula: C4H12Br2N2 Molecular Weight (g/mol): 247.962 InChI Key: UXWKNNJFYZFNDI-UHFFFAOYSA-N Synonym: Piperazinium Dibromide, Piperazinium Bromide PubChem CID: 3017157 IUPAC Name: piperazine;dihydrobromide SMILES: C1CNCCN1.Br.Br