accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 599-66-6

CH3SOOCH3

CAS RN 599-66-6

IUPAC Name: 1-methyl-4-(4-methylbenzenesulfonyl)benzene
Synonyms: 1-methyl-4-(4-methylbenzenesulfonyl)benzene
Molecular Weight (g/mol): 246.32
Molecular Formula: C14H14O2S
InChi Key: WEAYCYAIVOIUMG-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results