accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 599-67-7

CH3HO

CAS RN 599-67-7

IUPAC Name: 1,1-diphenylethan-1-ol
Synonyms: 1,1-diphenylethanol
Molecular Weight (g/mol): 198.27
Molecular Formula: C14H14O
InChi Key: GIMDPFBLSKQRNP-UHFFFAOYSA-N
SMILES: CC(O)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results