accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FBr

CAS RN 59907-13-0

IUPAC Name: 2-bromo-1-fluoro-3-methylbenzene
Synonyms: 2-bromo-1-fluoro-3-methylbenzene
Molecular Weight (g/mol): 189.03
Molecular Formula: C7H6BrF
InChi Key: FYCXRRYRNRDSRM-UHFFFAOYSA-N
SMILES: CC1=CC=CC(F)=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results