accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59937-28-9

IUPAC Name: 1,3-bis(propan-2-yl) 2-(2H-1,3-dithiol-2-ylidene)propanedioate
Synonyms: malotilate malotilatum 马洛替酯 kantec malotilato
Molecular Weight (g/mol): 288.38
Molecular Formula: C12H16O4S2
InChi Key: YPIQVCUJEKAZCP-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)C(C(=O)OC(C)C)=C1SC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results