accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNNOCl

CAS RN 59944-65-9

IUPAC Name: 4-methyl-1,2,3-thiadiazole-5-carbonyl chloride
Synonyms: 4-methyl-1,2,3-thiadiazole-5-carbonyl chloride
Molecular Weight (g/mol): 162.59
Molecular Formula: C4H3ClN2OS
InChi Key: SDNDOCTUXWLDIX-UHFFFAOYSA-N
SMILES: CC1=C(SN=N1)C(Cl)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results