accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 59944-76-2

IUPAC Name: thieno[2,3-b]pyridine-2-carboxylic acid
Synonyms: thieno[2,3-b]pyridine-2-carboxylic acid
Molecular Weight (g/mol): 179.19
Molecular Formula: C8H5NO2S
InChi Key: XGCSHAYMNOFFNA-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=CC=CN=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results