accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONCH2CCH2CH2

CAS RN 59990-69-1

IUPAC Name: (isocyanatomethyl)benzene; 1,4-diethenylbenzene; ethenylbenzene
Synonyms: benzene, 1,4-diethenyl-; benzyl isocyanate; styrene
Molecular Weight (g/mol): 367.49
Molecular Formula: C26H25NO
InChi Key: QMEHLMLNGYENTN-UHFFFAOYSA-N
SMILES: C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.O=C=NCC1=CC=CC=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)