accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NN(E/Z)

CAS RN 60-09-3

IUPAC Name: 4-(2-phenyldiazen-1-yl)aniline
Synonyms: 4-aminoazobenzene
Molecular Weight (g/mol): 197.24
Molecular Formula: C12H11N3
InChi Key: QPQKUYVSJWQSDY-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)N=NC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results