CAS RN 60-10-6
Dithizone, 98%
CAS: 60-10-6 Molecular Formula: C13H12N4S Molecular Weight (g/mol): 256.33 MDL Number: MFCD00003025 InChI Key: UOFGSWVZMUXXIY-UHFFFAOYSA-N Synonym: dithizone,dithizon,ditizon,carbazone, diphenylthio,usaf ek-3092,diazenecarbothioic acid, phenyl-, 2-phenylhydrazide,1,5-diphenyl-3-mercaptoformazan,3-formazanthiol, 1,5-diphenyl,unii-njz2cj4d6p,1,5-diphenyl-3-thiocarbazone PubChem CID: 657262 SMILES: S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
Dithizone, ACS, 85+%
CAS: 60-10-6 Molecular Formula: C13H12N4S Molecular Weight (g/mol): 256.33 MDL Number: MFCD00003025 InChI Key: UOFGSWVZMUXXIY-UHFFFAOYSA-N Synonym: dithizone,dithizon,ditizon,carbazone, diphenylthio,usaf ek-3092,diazenecarbothioic acid, phenyl-, 2-phenylhydrazide,1,5-diphenyl-3-mercaptoformazan,3-formazanthiol, 1,5-diphenyl,unii-njz2cj4d6p,1,5-diphenyl-3-thiocarbazone PubChem CID: 657262 IUPAC Name: 1-anilino-3-phenyliminothiourea SMILES: S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
Dithizone 85.0+%, TCI America™
CAS: 60-10-6 Molecular Formula: C13H12N4S Molecular Weight (g/mol): 256.33 MDL Number: MFCD00003025 InChI Key: UOFGSWVZMUXXIY-UHFFFAOYSA-N Synonym: dithizone,dithizon,ditizon,carbazone, diphenylthio,usaf ek-3092,diazenecarbothioic acid, phenyl-, 2-phenylhydrazide,1,5-diphenyl-3-mercaptoformazan,3-formazanthiol, 1,5-diphenyl,unii-njz2cj4d6p,1,5-diphenyl-3-thiocarbazone PubChem CID: 657262 IUPAC Name: 3-(phenylamino)-1-(phenylimino)thiourea SMILES: S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
Dithizone, ACS Reagent Grade, Ricca Chemical
CAS: 60-10-6 Molecular Formula: C13H12N4S Molecular Weight (g/mol): 256.33 MDL Number: MFCD00003025 InChI Key: UOFGSWVZMUXXIY-UHFFFAOYSA-N Synonym: dithizone,dithizon,ditizon,carbazone, diphenylthio,usaf ek-3092,diazenecarbothioic acid, phenyl-, 2-phenylhydrazide,1,5-diphenyl-3-mercaptoformazan,3-formazanthiol, 1,5-diphenyl,unii-njz2cj4d6p,1,5-diphenyl-3-thiocarbazone PubChem CID: 657262 IUPAC Name: 3-(phenylamino)-1-(phenylimino)thiourea SMILES: S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
Diphenylthiocarbazone, ACS Grade, LabChem™
CAS: 60-10-6 Molecular Formula: C13H12N4S Molecular Weight (g/mol): 256.33 MDL Number: MFCD00003025 InChI Key: UOFGSWVZMUXXIY-UHFFFAOYSA-N Synonym: dithizone,dithizon,ditizon,carbazone, diphenylthio,usaf ek-3092,diazenecarbothioic acid, phenyl-, 2-phenylhydrazide,1,5-diphenyl-3-mercaptoformazan,3-formazanthiol, 1,5-diphenyl,unii-njz2cj4d6p,1,5-diphenyl-3-thiocarbazone PubChem CID: 657262 IUPAC Name: 3-(phenylamino)-1-(phenylimino)thiourea SMILES: S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
Sigma Aldrich Dithizone
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 3,3'-DithiodIpr∗
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Dithizone
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Fisher Science Education™ Dithizone
CAS: 60-10-6 Molecular Formula: C13H12N4S Molecular Weight (g/mol): 256.33 MDL Number: MFCD00003025 InChI Key: UOFGSWVZMUXXIY-UHFFFAOYSA-N Synonym: dithizone,dithizon,ditizon,carbazone, diphenylthio,usaf ek-3092,diazenecarbothioic acid, phenyl-, 2-phenylhydrazide,1,5-diphenyl-3-mercaptoformazan,3-formazanthiol, 1,5-diphenyl,unii-njz2cj4d6p,1,5-diphenyl-3-thiocarbazone PubChem CID: 657262 IUPAC Name: 3-(phenylamino)-1-(phenylimino)thiourea SMILES: S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
Dithizone, MP Biomedicals
CAS: 60-10-6 Molecular Formula: C13H12N4S Molecular Weight (g/mol): 256.33 MDL Number: MFCD00003025 InChI Key: UOFGSWVZMUXXIY-UHFFFAOYSA-N Synonym: Diphenylthiocarbazone, Phenyldiazenecarbothioic acid 2-phenylhydrazide, (Phenylazo) thioformic acid 2-phenylhydrazide PubChem CID: 657262 IUPAC Name: 3-(phenylamino)-1-(phenylimino)thiourea SMILES: S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1