accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60-19-5

IUPAC Name: hydrogen 4-(2-aminoethyl)phenol chloride
Synonyms: hydrogen tyramine chloride
Molecular Weight (g/mol): 173.64
Molecular Formula: C8H12ClNO
InChi Key: RNISDHSYKZAWOK-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCC1=CC=C(O)C=C1
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (5)
  • (3)