accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60-25-3

IUPAC Name: trimethyl[6-(trimethylazaniumyl)hexyl]azanium dihydroxide
Synonyms: hexamethonium iodide hexamethonium tartrate
Molecular Weight (g/mol): 236.4
Molecular Formula: C12H32N2O2
InChi Key: GYLUMIIRFKDCKI-UHFFFAOYSA-L
SMILES: [OH-].[OH-].C[N+](C)(C)CCCCCC[N+](C)(C)C
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results