accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HN(S)CH3OHO

CAS RN 600-21-5

IUPAC Name: (2S)-2-(methylamino)propanoic acid
Synonyms: N-methylalanine
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: GDFAOVXKHJXLEI-VKHMYHEASA-N
SMILES: CN[C@@H](C)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)