accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 600-30-6

IUPAC Name: 2,3-dibromobutanoic acid
Synonyms: 2,3-dibromo-butyric acid
Molecular Weight (g/mol): 245.9
Molecular Formula: C4H6Br2O2
InChi Key: HESQKTULJLBDRF-UHFFFAOYNA-N
SMILES: CC(Br)C(Br)C(O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)