accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COHCH3H3C

CAS RN 600-36-2

IUPAC Name: 2,4-dimethylpentan-3-ol
Synonyms: 2,4-dimethyl-3-pentanol
Molecular Weight (g/mol): 116.2
Molecular Formula: C7H16O
InChi Key: BAYAKMPRFGNNFW-UHFFFAOYSA-N
SMILES: CC(C)C(O)C(C)C
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)